Learn More
enQuireBio™ Recombinant Human DUSP21 Protein
A cDNA sequence encoding the DUSP21 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11714-2ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
DUSP21 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMTASAS SFSSSQGVQQ PSIYSFSQIT RSLFLSNGVA ANDKLLLSSN RITAIVNASV EVVNVFFEGI QYIKVPVTDA RDSRLYDFFD PIADLIHTID MRQGRTLLHC MAGVSRSASL CLAYLMKYHS MSLLDAHTWT KSRRPIIRPN NGFWEQLINY EFKLFNNNTV RMINSPVGNI PDIYEKDLRM MISM | |
Greater than 90% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The DUSP21 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 200mM NaCl, 2mM DTT and 20% glycerol. |