missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Dynactin subunit 4 Protein
A DNA sequence encoding the Homo sapiens (Human) Dynactin subunit 4, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP9937-ye-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001129115.1 | |
Dynactin subunit 4 Protein | |
Research Use Only | |
DCTN4 | |
Recombinant Protein | |
Yeast | |
ASLLQSDRVL YLVQGEKKVR APLSQLYFCR YCSELRSLEC VSHEVDSHYC PSCLENMPSA EAKLKKNRCA NCFDCPGCMH TLSTRATSIS TQLPDDPAKT TMKKAYYLAC GFCRWTSRDV GMADKSVASG GWQEPENPHT QRMNKLIEYY QQLAQKEKVE RDRKKLARRR NYMPLAFSDK YGLGTRLQRP RAGASISTLA GLSLKEGEDQ KEIKIEPAQA VDEVEPLPED YYTRPVNLTE VTTLQQRLLQ PDFQPVCASQ LYPRHKHLLI KRSLRCRKCE HNLSKPEFNP TSIKFKIQLV AVNYIPEVRI MSIPNLRYMK ESQVLLTLTN PVENLTHVTL FECEEGDPDD INSTAKVVVP PKELVLAGKD AAAEYDELAE PQDFQDDPDI IAFRKANKVG IFIKVTPQRE EGEVTVCFKM KHDFKNLAAP IRPIEESDQG TEVIWLTQHV ELSLGPLLP | |
Greater than 90% as determined by SDS-PAGE. |
51164 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |