missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human E3 ubiquitin-protein ligase RBX1 Protein
A DNA sequence encoding the Homo sapiens (Human) E3 ubiquitin-protein ligase RBX1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6590-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_055063.1 | |
E3 ubiquitin-protein ligase RBX1 Protein | |
Research Use Only | |
RBX1 | |
Recombinant Protein | |
E. coli | |
MAAAMDVDTP SGTNSGAGKK RFEVKKWNAV ALWAWDIVVD NCAICRNHIM DLCIECQANQ ASATSEECTV AWGVCNHAFH FHCISRWLKT RQVCPLDNRE WEFQKYGH | |
Greater than 80% as determined by SDS-PAGE. |
9978 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |