missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human E3 ubiquitin-protein ligase ZNRF3 Protein
A DNA sequence encoding the Homo sapiens (Human) E3 ubiquitin-protein ligase ZNRF3, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8236-ec-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001193927.1 | |
E3 ubiquitin-protein ligase ZNRF3 Protein | |
Research Use Only | |
ZNRF3 | |
Recombinant Protein | |
E. coli | |
KETAFVEVVL FESSPSGDYT TYTTGLTGRF SRAGATLSAE GEIVQMHPLG LCNNNDEEDL YEYGWVGVVK LEQPELDPKP CLTVLGKAKR AVQRGATAVI FDVSENPEAI DQLNQGSEDP LKRPVVYVKG ADAIKLMNIV NKQKVARARI QHRPPRQPTE YFDM | |
Greater than 90% as determined by SDS-PAGE. |
84133 | |
500 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |