missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Elongation factor 1-delta Protein
A DNA sequence encoding the Homo sapiens (Human) Elongation factor 1-delta, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5960-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001123525.2 | |
Elongation factor 1-delta Protein | |
Research Use Only | |
EF1D | |
Recombinant Protein | |
E. coli | |
ATNFLAHEKI WFDKFKYDDA ERRFYEQMNG PVAGASRQEN GASVILRDIA RARENIQKSL AGSSGPGASS GTSGDHGELV VRIASLEVEN QSLRGVVQEL QQAISKLEAR LNVLEKSSPG HRATAPQTQH VSPMRQVEPP AKKPATPAED DEDDDIDLFG SDNEEEDKEA AQLREERLRQ YAEKKAKKPA LVAKSSILLD VKPWDDETDM AQLEACVRSI QLDGLVWGAS KLVPVGYGIR KLQIQCVVED DKVGTDLLEE EITKFEEHVQ SVDIAAFNKI | |
Greater than 90% as determined by SDS-PAGE. |
1936 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |