missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ENSA / Endosulfine alpha Protein
A DNA sequence encoding the Homo sapiens (Human) ENSA / Endosulfine alpha, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8383-ec-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_004427.1 | |
ENSA / Endosulfine alpha Protein | |
Research Use Only | |
ENSA | |
Recombinant Protein | |
E. coli | |
MSQKQEEENP AEETGEEKQD TQEKEGILPE RAEEAKLKAK YPSLGQKPGG SDFLMKRLQK GQKYFDSGDY NMAKAKMKNK QLPSAGPDKN LVTGDHIPTP QDLPQRKSSL VTSKLAGGQV E | |
Greater than 90% as determined by SDS-PAGE. |
2029 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |