missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Enterokinase Protein
A cDNA sequence encoding the Enterokinase was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11792-20IU
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
Enterokinase Protein | |
20 IU | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
50mM Tris-HCl, pH 8.0, 0.5M NaCl, 1mM CaCl2 and 50% glycerol. |
Greater than 95.0% as determined by HPLC-SEC. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSKRGISSR HHSLSSYEIM FAALFAILVV LCAGLIAVSC LTIKESQRGA ALGQSHEARA TFKITSGVTY NPNLQDKLSV DFKVLAFDLQ QMIDEIFLSS NLKNEYKNSR VLQFENGSII VVFDLFFAQW VSDENVKEEL IQGLEANKSS QLVTFHIDLN SVDILDKLTT TSHLATPGNV SIECLPGSSP CTDALTCIKA DLFCDGEVNC PDGSDEDNKM CATVCDGRFL LTGSSGSFQA THYPKPSETS VVCQWIIRVN QGLSIKLSFD DFNTYYTDIL DIYEGVGSSK ILRASIWETN PGTIRIFSNQ VTATFLIESD ESDYVGFNAT YTAFNSSELN NYEKINCNFE DGFCFWVQDL NDDNEWERIQ GSTFSPFTGP NFDHTFGNAS GFYISTPTGP GGRQERVGLL SLPLDPTLEP ACLSFWYHMY GENVHKLSIN ISNDQNMEKT VFQKEGNYGD NWNYGQVTLN ETVKFKVAFN AFKNKILSDI ALDDISLTYG ICNGSLYPEP TLVPTPPPEL PTDCGGPFEL WEPNTTFSST NFPNSYPNLA FCVWILNAQK GKNIQLHFQE FDLENINDVV EIRDGEEADS LLLAVYTGPG PVKDVFSTTN RMTVLLITND VLARGGFKAN FTTGYHLGIP EPCKADHFQC KNGECVPLVN LCDGHLHCED GSDEADCVRF FNGTTNNNGL VRFRIQSIWH TACAENWTTQ ISNDVCQLLG LGSGNSSKPI FPTDGGPFVK LNTAPDGHLI LTPSQQCLQD SLIRLQCNHK SCGKKLAAQD ITPKIVGGSN AKEGAWPWVV GLYYGGRLLC GASLVSSDWL VSAAHCVYGR NLEPSKWTAI LGLHMKSNLT SPQTVPRLID EIVINPHYNR RRKDNDIAMM HLEFKVNYTD YIQPICLPEE NQVFPPGRNC SIAGWGTVVY QGTTANILQE ADVPLLSNER CQQQMPEYNI TENMICAGYE EGGIDSCQGD SGGPLMCQEN NRWFLAGVTS FGYKCALPNR PGVYARVSRF TEWIQSFLH |