missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ERGIC3 Protein
A DNA sequence encoding the Homo sapiens (Human) ERGIC3, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8276-ec-200ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_057050.1 | |
ERGIC3 Protein | |
Research Use Only | |
ERGIC3 | |
Recombinant Protein | |
E. coli | |
QYYLTTEVHP ELYVDKSRGD KLKINIDVLF PHMPCAYLSI DAMDVAGEQQ LDVEHNLFKQ RLDKDGIPVS SEAERHELGK VEVTVFDPDS LDPDRCESCY GAEAEDIKCC NTCEDVREAY RRRGWAFKNP DTIEQCRREG FSQKMQEQKN EGCQVYGFLE VNKVAGNFHF APGKSFQQSH VHVHDLQSFG LDNINMTHYI QHLSFGEDYP GIVNPLDHTN VTAPQASMMF QYFVKVVPTV YMKVDGEVLR TNQFSVTRHE KVANGLLGDQ GLPGVFVLYE LSPMMVKLTE KHRSF | |
Greater than 90% as determined by SDS-PAGE. |
51614 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |