missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Erythropoietin / EPO Protein
A DNA sequence encoding the Human Erythropoietin / EPO, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10347-ma-15000IU
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_000790.2 | |
Erythropoietin / EPO Protein | |
Research Use Only | |
EPO | |
The ED50 determined by a cell proliferation assay using human megakaryoblastic leukemia cells is less than 2 ng/ml, corresponding to a specific activity of > 5.0 M 105 IU/mg. | |
Human | |
FC | |
Greater than 98% as determined by SDS-PAGE and HPLC. |
2056 | |
1500 IU | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
Mammal | |
0.2um filtered sodium citrate buffer (1 liter of ddH2O containing 5.9 g of sodium citrate, 5.8 g of sodium chloride and 0.06 g of citric acid), lyophilized |