missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Estrogen Receptor 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Estrogen Receptor 2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6000-ye-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001035365.1 | |
Estrogen Receptor 2 Protein | |
Research Use Only | |
ESR2 | |
Recombinant Protein | |
Yeast | |
DIKNSPSSLN SPSSYNCSQS ILPLEHGSIY IPSSYVDSHH EYPAMTFYSP AVMNYSIPSN VTNLEGGPGR QTTSPNVLWP TPGHLSPLVV HRQLSHLYAE PQKSPWCEAR SLEHTLPVNR ETLKRKVSGN RCASPVTGPG SKRDAHFCAV CSDYASGYHY GVWSCEGCKA FFKRSIQGHN DYICPATNQC TIDKNRRKSC QACRLRKCYE VGMVKCGSRR ERCGYRLVRR QRSADEQLHC AGKAKRSGGH APRVRELLLD ALSPEQLVLT LLEAEPPHVL ISRPSAPFTE ASMMMSLTKL ADKELVHMIS WAKKIPGFVE LSLFDQVRLL ESCWMEVLMM GLMWRSIDHP GKLIFAPDLV LDRDEGKCVE GILEIFDMLL ATTSRFRELK LQHKEYLCVK AMILLNSSMY PLVTATQDAD SSRKLAHLLN AVTDALVWVI AKSGISSQQQ SMRLANLLML LSHVRHASNK GMEHLLNMKC KNVVPVYDLL LEMLNAHVLR GCKSSITGSE CSPAEDSKSK EGSQNPQSQ | |
Greater than 90% as determined by SDS-PAGE. |
2100 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |