missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human F-box only protein 1
A DNA sequence encoding the Homo sapiens (Human) F-box only protein 3, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP9935-ye-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_036307.2 | |
F-box only protein 1 | |
Research Use Only | |
FBXO3 | |
Recombinant Protein | |
Yeast | |
MAAMETETAP LTLESLPTDP LLLILSFLDY RDLINCCYVS RRLSQLSSHD PLWRRHCKKY WLISEEEKTQ KNQCWKSLFI DTYSDVGRYI DHYAAIKKAW DDLKKYLEPR CPRMVLSLKE GAREEDLDAV EAQIGCKLPD DYRCSYRIHN GQKLVVPGLL GSMALSNHYR SEDLLDVDTA AGGFQQRQGL KYCLPLTFCI HTGLSQYIAV EAAEGRNKNE VFYQCPDQMA RNPAAIDMFI IGATFTDWFT SYVKNVVSGG FPIIRDQIFR YVHDPECVAT TGDITVSVST SFLPELSSVH PPHYFFTYRI RIEMSKDALP EKACQLDSRY WRITNAKGDV EEVQGPGVVG EFPIISPGRV YEYTSCTTFS TTSGYMEGYY TFHFLYFKDK IFNVAIPRFH MACPTFRVSI ARLEMGPDEY EEMEEEEEEE EEEDEDDDSA DMDESDEDDE EERRRRVFDV PIRRRRCSRL F | |
Greater than 90% as determined by SDS-PAGE. |
26273 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |