missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human F-box / WD repeat-containing protein 1
A DNA sequence encoding the Homo sapiens (Human) F-box/WD repeat-containing protein 4, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6025-ec-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_071322.1 | |
F-box / WD repeat-containing protein 1 | |
Research Use Only | |
FBXW4 | |
Recombinant Protein | |
E. coli | |
MAAAAGEEEE EEEAARESAA RPAAGPALWR LPEELLLLIC SYLDMRALGR LAQVCRWLRR FTSCDLLWRR IARASLNSGF TRLGTDLMTS VPVKERVKVS QNWRLGRCRE GILLKWRCSQ MPWMQLEDDS LYISQANFIL AYQFRPDGAS LNRRPLGVFA GHDEDVCHFV LANSHIVSAG GDGKIGIHKI HSTFTVKYSA HEQEVNCVDC KGGIIVSGSR DRTAKVWPLA SGRLGQCLHT IQTEDRVWSI AISPLLSSFV TGTACCGHFS PLRIWDLNSG QLMTHLGSDF PPGAGVLDVM YESPFTLLSC GYDTYVRYWD LRTSVRKCVM EWEEPHDSTL YCLQTDGNHL LATGSSYYGV VRLWDRRQRA CLHAFPLTST PLSSPVYCLR LTTKHLYAAL SYNLHVLDFQ NP | |
Greater than 90% as determined by SDS-PAGE. |
6468 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |