missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human FABP1 Protein

A cDNA sequence encoding the FABP1 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP11832-25ug

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 15948518

  • 181.00€ / 25µg

Spedizione stimata: 07-02-2023
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

Specifica

Specifica

FABP1 Protein
25 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The protein solution (1 mg/ml) contains 20mM Tris-HCl pH 8.
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MSFSGKYQLQ SQENFEAFMK AIGLPEELIQ KGKDIKGVSEIVQNGKHFKF TITAGSKVIQ NEFTVGEECE LETMTGEKVK TVVQLEGDNK LVTTFKNIKSVTELNGDIIT NTMTLGDIVF KRISKRI
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali