missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FAP / Seprase Protein
A DNA sequence encoding the Homo sapiens (Human) FAP / Seprase, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6020-ec-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001278736.1 | |
FAP / Seprase Protein | |
Research Use Only | |
FAP | |
Recombinant Protein | |
E. coli | |
LRPSRVHNSE ENTMRALTLK DILNGTFSYK TFFPNWISGQ EYLHQSADNN IVLYNIETGQ SYTILSNRTM KSVNASNYGL SPDRQFVYLE SDYSKLWRYS YTATYYIYDL SNGEFVRGNE LPRPIQYLCW SPVGSKLAYV YQNNIYLKQR PGDPPFQITF NGRENKIFNG IPDWVYEEEM LATKYALWWS PNGKFLAYAE FNDTDIPVIA YSYYGDEQYP RTINIPYPKA GAKNPVVRIF IIDTTYPAYV GPQEVPVPAM IASSDYYFSW LTWVTDERVC LQWLKRVQNV SVLSICDFRE DWQTWDCPKT QEHIEESRTG WAGGFFVSTP VFSYDAISYY KIFSDKDGYK HIHYIKDTVE NAIQITSGKW EAINIFRVTQ DSLFYSSNEF EEYPGRRNIY RISIGSYPPS KKCVTCHLRK ERCQYYTASF SDYAKYYALV CYGPGIPIST LHDGRTDQEI KILEENKELE NALKNIQLPK EEIKKLEVDE ITLWYKMILP PQFDRSKKYP LLIQVYGGPC SQSVRSVFAV NWISYLASKE GMVIALVDGR GTAFQGDKLL YAVYRKLGVY EVEDQITAVR KFIEMGFIDE KRIAIWGWSY GGYVSSLALA SGTGLFKCGI AVAPVSSWEY YASVYTERFM GLPTKDDNLE HYKNSTVMAR AEYFRNVDYL LIHGTADDNV HFQNSAQIAK ALVNAQVDFQ AMWYSDQNHG LSGLSTNHLY THMTHFLKQC FSLSD | |
Greater than 90% as determined by SDS-PAGE. |
2191 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |