missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FK506-binding protein-like Protein
A DNA sequence encoding the Homo sapiens (Human) FK506-binding protein-like, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8189-ec-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_071393.2 | |
FK506-binding protein-like Protein | |
Research Use Only | |
FKBPL | |
Recombinant Protein | |
E. coli | |
METPPVNTIG EKDTSQPQQE WEKNLRENLD SVIQIRQQPR DPPTETLELE VSPDPASQIL EHTQGAEKLV AELEGDSHKS HGSTSQMPEA LQASDLWYCP DGSFVKKIVI RGHGLDKPKL GSCCRVLALG FPFGSGPPEG WTELTMGVGP WREETWGELI EKCLESMCQG EEAELQLPGH SGPPVRLTLA SFTQGRDSWE LETSEKEALA REERARGTEL FRAGNPEGAA RCYGRALRLL LTLPPPGPPE RTVLHANLAA CQLLLGQPQL AAQSCDRVLE REPGHLKALY RRGVAQAALG NLEKATADLK KVLAIDPKNR AAQEELGKVV IQGKNQDAGL AQGLRKMFG | |
Greater than 80% as determined by SDS-PAGE. |
63943 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |