missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FLT1 Protein
A cDNA sequence encoding the FLT1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10611-100ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
2321 | |
100 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The activity of FLT1 was determined by its ability to inhibit the VEGF(165)-induced proliferation of HUVECs. | |
Human | |
Untagged | |
FLT1 was lyophilized from a concentrated (1 mg/ml) sterile solution containing 1xPBS. |
FLT1 Protein | |
Research Use Only | |
FLT1 | |
Recombinant Protein | |
Baculovirus | |
MVSYWDTGVL LCALLSCLLL TGSSSGSKLK DPELSLKGTQ HIMQAGQTLH LQCRGEAAHK WSLPEMVSKE SERLSITKSA CGRNGKQFCS TLTLNTAQAN HTGFYSCKYL AVPTSKKKET ESAIYIFISD TGRPFVEMYS EIPEIIHMTE GRELVIPCRV TSPNITVTLK KFPLDTLIPD GKRIIWDSRK GFIISNATYK EIGLLTCEAT VNGHLYKTNY LTHRQTNTII DVQISTPRPV KLLRGHTLVL NCTATTPLNT RVQMTWSYPD EKNKRASVRR RIDQSNSHAN IFYSVLTIDK MQNKDKGLYT CRVRSGPSFK SVNTSVHIYD KAFITVKHRK QQVLETVAGK RSYRLSMKVK AFPSPEVVWL KDGLPATEKS ARYLTRGYSL IIKDVTEEDA GNYTILLSIK QSNVFKNLTA TLIVNVKPQI YEKAVSSFPD PALYPLGSRQ ILTCTAYGIP QPTIKWFWHP CNHNHSEARC DFCSNNEESF ILDADSNMGN RIESITQRMA IIEGKNKMAS TLVVADSRIS GIYICIASNK VGTVGRNISF YITDVPNGFH VNLEKMPTEG EDLKLSCTVN KFLYRDVTWI LLRTVNNRTM HYSISKQKMA ITKEHSITLN LTIMNVSLQD SGTYACRARN VYTGEEILQK KEITIRGEHC NKKAVFSRIS KFKSTRNDCT TQSNVKH | |
Greater than 95.0% as determined by SDS-PAGE. |