missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Fructose-1,6-bisphosphatase 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Fructose-1,6-bisphosphatase 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6023-ec-100ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_000498.2 | |
Fructose-1,6-bisphosphatase 1 Protein | |
Research Use Only | |
FBP1 | |
Recombinant Protein | |
E. coli | |
ADQAPFDTDV NTLTRFVMEE GRKARGTGEL TQLLNSLCTA VKAISSAVRK AGIAHLYGIA GSTNVTGDQV KKLDVLSNDL VMNMLKSSFA TCVLVSEEDK HAIIVEPEKR GKYVVCFDPL DGSSNIDCLV SVGTIFGIYR KKSTDEPSEK DALQPGRNLV AAGYALYGSA TMLVLAMDCG VNCFMLDPAI GEFILVDKDV KIKKKGKIYS LNEGYARDFD PAVTEYIQRK KFPPDNSAPY GARYVGSMVA DVHRTLVYGG IFLYPANKKS PNGKLRLLYE CNPMAYVMEK AGGMATTGKE AVLDVIPTDI HQRAPVILGS PDDVLEFLKV YEKHSAQ | |
Greater than 80% as determined by SDS-PAGE. |
2203 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |