missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Galectin-1 / LGALS1 Protein
A DNA sequence encoding the Human Galectin-1 / LGALS1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10191-ec-250ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_002296.1 | |
Galectin-1 / LGALS1 Protein | |
Research Use Only | |
LGALS1 | |
The biological activity determined by a chemotaxis bioassay using human blood monocytes is in a concentration range of 1.0-10 ng/ml. | |
Human | |
Untagged | |
0.2 um filtered 20 mM PBS, pH 7.4, 150mM NaCl, lyophilized |
3956 | |
250 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
ACGLVASNLN LKPGECLRVR GEVAPDAKSF VLNLGKDSNN LCLHFNPRFN AHGDANTIVC NSKDGGAWGT EQREAVFPFQ PGSVAEVCIT FDQANLTVKL PDGYEFKFPN RLNLEAINYM AADGDFKIKC VAFD | |
Greater than 95% as determined by SDS-PAGE and HPLC. |