missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GALNT14 Protein
A DNA sequence encoding the Homo sapiens (Human) GALNT14, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7871-ec-200ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001240755.1 | |
GALNT14 Protein | |
Research Use Only | |
GALNT14 | |
Recombinant Protein | |
E. coli | |
MRRLTRRLVL PVFGVLWITV LLFFWVTKRK LEVPTGPEVQ TPKPSDADWD DLWDQFDERR YLNAKKWRVG DDPYKLYAFN QRESERISSN RAIPDTRHLR CTLLVYCTDL PPTSIIITFH NEARSTLLRT IRSVLNRTPT HLIREIILVD DFSNDPDDCK QLIKLPKVKC LRNNERQGLV RSRIRGADIA QGTTLTFLDS HCEVNRDWLQ PLLHRVKEDY TRVVCPVIDI INLDTFTYIE SASELRGGFD WSLHFQWEQL SPEQKARRLD PTEPIRTPII AGGLFVIDKA WFDYLGKYDM DMDIWGGENF EISFRVWMCG GSLEIVPCSR VGHVFRKKHP YVFPDGNANT YIKNTKRTAE VWMDEYKQYY YAARPFALER PFGNVESRLD LRKNLRCQSF KWYLENIYPE LSIPKESSIQ KGNIRQRQKC LESQRQNNQE TPNLKLSPCA KVKGEDAKSQ VWAFTYTQQI LQEELCLSVI TLFPGAPVVL VLCKNGDDRQ QWTKTGSHIE HIASHLCLDT DMFGDGTENG KEIVVNPCES SLMSQHWDMV SS | |
Greater than 90% as determined by SDS-PAGE. |
79623 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |