missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GATC Protein
A DNA sequence encoding the Homo sapiens (Human) GATC, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6071-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_789788.1 | |
GATC Protein | |
Research Use Only | |
GATC | |
Recombinant Protein | |
E. coli | |
MWSRLVWLGL RAPLGGRQGF TSKADPQGSG RITAAVIEHL ERLALVDFGS REAVARLEKA IAFADRLRAV DTDGVEPMES VLEDRCLYLR SDNVVEGNCA DELLQNSHRV VEEYFVAPPG NISLPKLDEQ EPFPHS | |
Greater than 80% as determined by SDS-PAGE. |
283459 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |