missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GDF5 Protein
Recombinant Protein
Marca: enQuireBio™ QP5375-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_000548.2 | |
GDF5 Protein | |
Research Use Only | |
GDF5 | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |
8200 | |
50 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MAPLATRQGK RPSKNLKARC SRKALHVNFK DMGWDDWIIA PLEYEAFHCE GLCEFPLRSH LEPTNHAVIQ TLMNSMDPES TPPTCCVPTR LSPISILFID SANNVVYKQY EDMVVESCGC R | |
Greater than 95% as determined by SDS-PAGE. |