missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Gem-associated protein 1
A DNA sequence encoding the Homo sapiens (Human) Gem-associated protein 7, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8052-ec-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001007270.1 | |
Gem-associated protein 1 | |
Research Use Only | |
GEMIN7 | |
Recombinant Protein | |
E. coli | |
MQTPVNIPVP VLRLPRGPDG FSRGFAPDGR RAPLRPEVPE IQECPIAQES LESQEQRARA ALRERYLRSL LAMVGHQVSF TLHEGVRVAA HFGATDLDVA NFYVSQLQTP IGVQAEALLR CSDIISYTFK P | |
Greater than 90% as determined by SDS-PAGE. |
79760 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |