missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GGPS1 / GGPPS1 Protein
A DNA sequence encoding the Homo sapiens (Human) GGPS1 / GGPPS1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6082-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001032354.1 | |
GGPS1 / GGPPS1 Protein | |
Research Use Only | |
GGPS1 | |
Recombinant Protein | |
E. coli | |
MEKTQETVQR ILLEPYKYLL QLPGKQVRTK LSQAFNHWLK VPEDKLQIII EVTEMLHNAS LLIDDIEDNS KLRRGFPVAH SIYGIPSVIN SANYVYFLGL EKVLTLDHPD AVKLFTRQLL ELHQGQGLDI YWRDNYTCPT EEEYKAMVLQ KTGGLFGLAV GLMQLFSDYK EDLKPLLNTL GLFFQIRDDY ANLHSKEYSE NKSFCEDLTE GKFSFPTIHA IWSRPESTQV QNILRQRTEN IDIKKYCVHY LEDVGSFEYT RNTLKELEAK AYKQIDARGG NPELVALVKH LSKMFKEENE | |
Greater than 80.8% as determined by SDS-PAGE. |
9453 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |