missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GLIPR1 / RTVP1 Protein
A DNA sequence encoding the Homo sapiens (Human) GLIPR1 / RTVP1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6089-ec-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_006842.2 | |
GLIPR1 / RTVP1 Protein | |
Research Use Only | |
GLIPR1 | |
Recombinant Protein | |
E. coli | |
ANILPDIENE DFIKDCVRIH NKFRSEVKPT ASDMLYMTWD PALAQIAKAW ASNCQFSHNT RLKPPHKLHP NFTSLGENIW TGSVPIFSVS SAITNWYDEI QDYDFKTRIC KKVCGHYTQV VWADSYKVGC AVQFCPKVSG FDALSNGAHF ICNYGPGGNY PTWPYKRGAT CSACPNNDKC LDNLCVNRQR DQVKRYYSVV YPGWPIYPRN R | |
Greater than 90% as determined by SDS-PAGE. |
11010 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |