missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GLP-1R / GLP1R Protein
A DNA sequence encoding the Homo sapiens (Human) GLP-1R / GLP1R, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6090-ec-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_002053.3 | |
GLP-1R / GLP1R Protein | |
Research Use Only | |
Glp1r | |
Recombinant Protein | |
E. coli | |
RPQGATVSLW ETVQKWREYR RQCQRSLTED PPPATDLFCN RTFDEYACWP DGEPGSFVNV SCPWYLPWAS SVPQGHVYRF CTAEGLWLQK DNSSLPWRDL SECEESKRGE RSSPEEQLLF LY | |
Greater than 98.9% as determined by SDS-PAGE. |
2740 | |
10 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |