missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Glucosamine-6-phosphate isomerase 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Glucosamine-6-phosphate isomerase 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6104-ec-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_005462.1 | |
Glucosamine-6-phosphate isomerase 1 Protein | |
Research Use Only | |
GNPDA1 | |
Recombinant Protein | |
E. coli | |
MKLIILEHYS QASEWAAKYI RNRIIQFNPG PEKYFTLGLP TGSTPLGCYK KLIEYYKNGD LSFKYVKTFN MDEYVGLPRD HPESYHSFMW NNFFKHIDIH PENTHILDGN AVDLQAECDA FEEKIKAAGG IELFVGGIGP DGHIAFNEPG SSLVSRTRVK TLAMDTILAN ARFFDGELTK VPTMALTVGV GTVMDAREVM ILITGAHKAF ALYKAIEEGV NHMWTVSAFQ QHPRTVFVCD EDATLELKVK TVKYFKGLML VHNKLVDPLY SIKEKETEKS QSSKKPYSD | |
Greater than 90% as determined by SDS-PAGE. |
10007 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |