missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Glutaredoxin-2, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) Glutaredoxin-2, mitochondrial, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8154-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001230328.1 | |
Glutaredoxin-2, mitochondrial Protein | |
Research Use Only | |
GLRX2 | |
Recombinant Protein | |
E. coli | |
MESNTSSSLE NLATAPVNQI QETISDNCVV IFSKTSCSYC TMAKKLFHDM NVNYKVVELD LLEYGNQFQD ALYKMTGERT VPRIFVNGTF IGGATDTHRL HKEGKLLPLV HQCYLKKSKR KEFQ | |
Greater than 80% as determined by SDS-PAGE. |
51022 | |
50 μg | |
Not determined. | |
Full Length of BC028113 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |