missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Glycine N-acyltransferase-like protein 1
A DNA sequence encoding the Homo sapiens (Human) Glycine N-acyltransferase-like protein 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7893-ec-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001207423.1 | |
Glycine N-acyltransferase-like protein 1 | |
Research Use Only | |
GLYATL1 | |
Recombinant Protein | |
E. coli | |
MILLNNSHKL LALYKSLARS IPESLKVYGS VYHINHGNPF NMEVLVDSWP EYQMVIIRPQ KQEMTDDMDS YTNVYRMFSK EPQKSEEVLK NCEIVNWKQR LQIQGLQESL GEGIRVATFS KSVKVEHSRA LLLVTEDILK LNASSKSKLG SWAETGHPDD EFESETPNFK YAQLDVSYSG LVNDNWKRGK NERSLHYIKR CIEDLPAACM LGPEGVPVSW VTMDPSCEVG MAYSMEKYRR TGNMARVMVR YMKYLRQKNI PFYISVLEEN EDSRRFVGQF GFFEASCEWH QWTCYPQNLV PF | |
Greater than 80% as determined by SDS-PAGE. |
92292 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |