missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GPC4 Protein
A cDNA sequence encoding the GPC4 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12034-2ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
GPC4 Protein | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
GPC4 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl and 10% glycerol. |
Greater than 85% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSSSLP SNVCNDERMA AGNGNEDDCW NGKGKSRYLF AVTGNGLANQ GNNPEVQVDT SKPDILILRQ IMALRVMTSK MKNAYNGNDV DFFDISDESS GEGSGSGCEY QQCPSEFDYN ATDHAGKSAN EKADS |