missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GrpE protein homolog 1, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) GrpE protein homolog 1, mitochondrial, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8144-ec-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_079472.1 | |
GrpE protein homolog 1, mitochondrial Protein | |
Research Use Only | |
GRPEL1 | |
Recombinant Protein | |
E. coli | |
CTATKQKNSG QNLEEDMGQS EQKADPPATE KTLLEEKVKL EEQLKETVEK YKRALADTEN LRQRSQKLVE EAKLYGIQAF CKDLLEVADV LEKATQCVPK EEIKDDNPHL KNLYEGLVMT EVQIQKVFTK HGLLKLNPVG AKFDPYEHEA LFHTPVEGKE PGTVALVSKV GYKLHGRTLR PALVGVVKEA | |
Greater than 80% as determined by SDS-PAGE. |
80273 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |