missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Hemoglobin subunit zeta Protein
A DNA sequence encoding the Homo sapiens (Human) Hemoglobin subunit zeta, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6148-ec-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_005323.1 | |
Hemoglobin subunit zeta Protein | |
Research Use Only | |
HBZ | |
Recombinant Protein | |
E. coli | |
SLTKTERTII VSMWAKISTQ ADTIGTETLE RLFLSHPQTK TYFPHFDLHP GSAQLRAHGS KVVAAVGDAV KSIDDIGGAL SKLSELHAYI LRVDPVNFKL LSHCLLVTLA ARFPADFTAE AHAAWDKFLS VVSSVLTEKY R | |
Greater than 80% as determined by SDS-PAGE. |
3050 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |