missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Heterogeneous nuclear ribonucleoprotein 1
A DNA sequence encoding the Homo sapiens (Human) Heterogeneous nuclear ribonucleoprotein L, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6176-ec-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001005335.1 | |
Heterogeneous nuclear ribonucleoprotein 1 | |
Research Use Only | |
HNRNPL | |
Recombinant Protein | |
E. coli | |
GENYDDPHKT PASPVVHIRG LIDGVVEADL VEALQEFGPI SYVVVMPKKR QALVEFEDVL GACNAVNYAA DNQIYIAGHP AFVNYSTSQK ISRPGDSDDS RSVNSVLLFT ILNPIYSITT DVLYTICNPC GPVQRIVIFR KNGVQAMVEF DSVQSAQRAK ASLNGADIYS GCCTLKIEYA KPTRLNVFKN DQDTWDYTNP NLSGQGDPGS NPNKRQRQPP LLGDHPAEYG GPHGGYHSHY HDEGYGP | |
Greater than 90% as determined by SDS-PAGE. |
3191 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |