missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HIF-1 alpha / HIF1A Protein
A DNA sequence encoding the Homo sapiens (Human) HIF-1 alpha / HIF1A, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP1121-ec-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001230013.1 | |
HIF-1 alpha / HIF1A Protein | |
Research Use Only | |
HIF1A | |
Recombinant Protein | |
E. coli | |
QLSPLESSSA SPESASPQST VTVFQQTQIQ EPTANATTTT ATTDELKTVT KDRMEDIKIL IASPSPTHIH KETTSATSSP YRDTQSRTAS PNRAGKGVIE QTEKSHPRSP NVLSVALSQR TTVPEEELNP KILALQNAQR KRKMEHDGSL FQAVGIGTLL QQPDDHAATT SLSWKRVKGC KSSEQNGMEQ KTIILIPSDL ACRLLGQSMD ESGLPQLTSY DCEVNAPIQG SRNLLQGEEL LRALDQVN | |
Greater than 90% as determined by SDS-PAGE. |
3091 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |