missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human High affinity copper uptake protein 1
A DNA sequence encoding the Homo sapiens (Human) High affinity copper uptake protein 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP9957-iv-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001850.1 | |
High affinity copper uptake protein 1 | |
Research Use Only | |
SLC31A1 | |
Recombinant Protein | |
E. coli | |
MDHSHHMGMS YMDSNSTMQP SHHHPTTSAS HSHGGGDSSM MMMPMTFYFG FKNVELLFSG LVINTAGEMA GAFVAVFLLA MFYEGLKIAR ESLLRKSQVS IRYNSMPVPG PNGTILMETH KTVGQQMLSF PHLLQTVLHI IQVVISYFLM LIFMTYNGYL CIAVAAGAGT GYFLFSWKKA VVVDITEHCH | |
Greater than 85% as determined by SDS-PAGE. |
1317 | |
50 μg | |
Not determined. | |
Full Length Transmembrane Protein | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |