missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Histone H2A.J Protein
A DNA sequence encoding the Homo sapiens (Human) Histone H2A.J, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6138-ec-200ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_808760.1 | |
Histone H2A.J Protein | |
Research Use Only | |
H2AFJ | |
Recombinant Protein | |
E. coli | |
SGRGKQGGKV RAKAKSRSSR AGLQFPVGRV HRLLRKGNYA ERVGAGAPVY LAAVLEYLTA EILELAGNAA RDNKKTRIIP RHLQLAIRND EELNKLLGKV TIAQGGVLPN IQAVLLPKKT ESQKTKSK | |
Greater than 90% as determined by SDS-PAGE. |
55766 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |