missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HIV-1 (group M, subtype A, isolate HXB2) Envelope glycoprotein gp160 Protein
A DNA sequence encoding the Homo sapiens (Human) HIV-1 (group M, subtype A, isolate HXB2) Envelope glycoprotein gp160, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7356-ec-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_057856.1 | |
HIV-1 (group M, subtype A, isolate HXB2) Envelope glycoprotein gp160 Protein | |
Research Use Only | |
gp120 | |
Recombinant Protein | |
E. coli | |
KLWVTVYYGV PVWKEATTTL FCASDAKAYD TEVHNVWATH ACVPTDPNPQ EVVLVNVTEN FNMWKNDMVE QMHEDIISLW DQSLKPCVKL TPLCVSLKCT DLKNDTNTNS SSGRMIMEKG EIKNCSFNIS TSIRGKVQKE YAFFYKLDII PIDNDTTSYK LTSCNTSVIT QACPKVSFEP IPIHYCAPAG FAILKCNNKT FNGTGPCTNV STVQCTHGIR PVVSTQLLLN GSLAEEEVVI RSVNFTDNAK TIIVQLNTSV EINCTRPNNN TRKRIRIQRG PGRAFVTIGK IGNMRQAHCN ISRAKWNNTL KQIASKLREQ FGNNKTIIFK QSSGGDPEIV THSFNCGGEF FYCNSTQLFN STWFNSTWST EGSNNTEGSD TITLPCRIKQ IINMWQKVGK AMYAPPISGQ IRCSSNITGL LLTRDGGNSN NESEIFRPGG GDMRDNWRSE LYKYKVVKIE PLGVAPTKAK RRVVQREKR | |
Greater than 90% as determined by SDS-PAGE. |
155971 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |