missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Homeobox protein MOX-1 Protein
A DNA sequence encoding the Homo sapiens (Human) Homeobox protein MOX-1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6354-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001035091.1 | |
Homeobox protein MOX-1 Protein | |
Research Use Only | |
MEOX1 | |
Recombinant Protein | |
E. coli | |
MDPAASSCMR SLQPPAPVWG CLRNPHSEGN GASGLPHYPP TPFSFHQKPD FLATATAAYP DFSASCLAAT PHSLPQEEHI FTEQHPAFPQ SPNWHFPVSD ARRRPNSGPA GGSKEMGTSS LGLVDTTGGP GDDYGVLGST ANETEKKSSR RRKESSDNQE NRGKPEGSSK ARKERTAFTK EQLRELEAEF AHHNYLTRLR RYEIAVNLDL SERQVKVWFQ NRRMKWKRVK GGQPISPNGQ DPEDGDSTAS PSSE | |
Greater than 80% as determined by SDS-PAGE. |
4222 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |