missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Homeobox protein OTX2 Protein
A DNA sequence encoding the Homo sapiens (Human) Homeobox protein OTX2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6458-ec-200ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001257452.1 | |
Homeobox protein OTX2 Protein | |
Research Use Only | |
OTX2 | |
Recombinant Protein | |
E. coli | |
MMSYLKQPPY AVNGLSLTTS GMDLLHPSVG YPGPWASCPA ATPRKQRRER TTFTRAQLDV LEALFAKTRY PDIFMREEVA LKINLPESRV QVWFKNRRAK CRQQQQQQQN GGQNKVRPAK KKTSPAREVS SESGTSGQFT PPSSTSVPTI ASSSAPVSIW SPASISPLSD PLSTSSSCMQ RSYPMTYTQA SGYSQGYAGS TSYFGGMDCG SYLTPMHHQL PGPGATLSPM GTNAVTSHLN QSPASLSTQG YGASSLGFNS TTDCLDYKDQ TASWKLNFNA DCLDYKDQTS SWKFQVL | |
Greater than 90% as determined by SDS-PAGE. |
5015 | |
200 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |