missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IFN a 1 Protein
A cDNA sequence encoding the IFN a 1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12373-5ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
IFN a 1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MCDLPETHSL DNRRTLMLLA QMSRISPSSC LMDRHDFGFP QEEFDGNQFQ KAPAISVLHE LIQQIFNLFT TKDSSAAWDE DLLDKFCTEL YQQLNDLEAC VMQEERVGET PLMNVDSILA VKKYFRRITL YLTEKKYSPC AWEVVRAEIM RSLSLSTNLQ ERLRRKE | |
Greater than 95.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
1 mg/ml in phosphate-buffered saline (PBS), pH 7.4. |