missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IGF1 Isoform 2 Protein
A DNA sequence encoding the Human IGF1 Isoform 2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10390-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_000609.1 | |
IGF1 Isoform 2 Protein | |
Research Use Only | |
IGF1 | |
The ED50 as determined by a cell proliferation assay using serum free human MCF-7 cells is less than 2 ng/ml, corresponding to a specific activity of > 5.0 M 105 IU/mg. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.0, lyophilized |
3479 | |
50 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
GPETLCGAEL VDALQFVCGD RGFYFNKPTG YGSSSRRAPQ TGIVDECCFR SCDLRRLEMY CAPLKPAKSA | |
Greater than 97% as determined by SDS-PAGE and HPLC. |