missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IGLL1 Protein
A cDNA sequence encoding the IGLL1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12388-20ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
3543 | |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSLLRPTAA SQSRALGPGA PGGSSRSSLR SRWGRFLLQR GSWTGPRCWP RGFQSKHNSV THVFGSGTQL TVLSQPKATP SVTLFPPSSE ELQANKATLV CLMNDFYPGI LTVTWKADGT PITQGVEMTT PSKQSNNKYA ASSYLSLTPE QWRSRRSYSC QVMHEGSTVE KTVAPAECS | |
Greater than 85% as determined by SDS-PAGE. |
IGLL1 Protein | |
Research Use Only | |
IGLL1 | |
Human | |
His | |
IGLL1 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M urea and 10% glycerol. |