missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL-11 / interleukin 11 Protein
A DNA sequence encoding the Human IL-11 / interleukin 11, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10319-ec-100ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_000632.1 | |
IL-11 / interleukin 11 Protein | |
Research Use Only | |
IL11 | |
The ED50 as determined by a cell proliferation assay using murine B9-11 cells is less than 1 ng/ml, corresponding to a specific activity of > 1.0 M 106 IU/mg. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
3589 | |
100 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
MPGPPPGPPR VSPDPRAELD STVLLTRSLL ADTRQLAAQL RDKFPADGDH NLDSLPTLAM SAGALGALQL PGVLTRLRAD LLSYLRHVQW LRRAGGSSLK TLEPELGTLQ ARLDRLLRRL QLLMSRLALP QPPPDPPAPP LAPPSSAWGG IRAAHAILGG LHLTLDWAVR GLLLLKTRL | |
Greater than 95% as determined by SDS-PAGE and HPLC. |