Learn More
enQuireBio™ Recombinant Human IL33 / Interleukin-33 / NF-HEV Protein
Recombinant Protein
Marca:  enQuireBio™ QP1168-1 mg
Dettagli aggiuntivi : Peso : 0.01000kg
Codice prodotto. 15938018
Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.
Specifica
NP_001186569.1
IL33 / Interleukin-33 / NF-HEV Protein
Research Use Only
IL33
Recombinant Protein
E. coli
MSITGISPIT EYLASLSTYN DQSITFALED ESYEIYVEDL KKDEKKDKVL LSYYESQHPS NESGDGVDGK MLMVTLSPTK DFWLHANNKE HSVELHKCEK PLPDQAFFVL HNMHSNCVSF ECKTDPGVFI GVKDNHLALI KVDSSENLCT ENILFKLSET
Greater than 95% as determined by SDS-PAGE.
90865
1 mg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Shows bioactivity at a concentration of 500 pg/mL in the D10S cell proliferation assay. The bioactivity acceptance criteria is set at 2 x 10^6 units/mg.
Human
Untagged
10 mM sodium phosphate, pH 7.5