missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL33 / Interleukin-33 / NF-HEV Protein
Recombinant Protein
Marca: enQuireBio™ QP1168-100 ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001186569.1 | |
IL33 / Interleukin-33 / NF-HEV Protein | |
Research Use Only | |
IL33 | |
Recombinant Protein | |
E. coli | |
MSITGISPIT EYLASLSTYN DQSITFALED ESYEIYVEDL KKDEKKDKVL LSYYESQHPS NESGDGVDGK MLMVTLSPTK DFWLHANNKE HSVELHKCEK PLPDQAFFVL HNMHSNCVSF ECKTDPGVFI GVKDNHLALI KVDSSENLCT ENILFKLSET | |
Greater than 95% as determined by SDS-PAGE. |
90865 | |
100 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity at a concentration of 500 pg/mL in the D10S cell proliferation assay. The bioactivity acceptance criteria is set at 2 x 10^6 units/mg. | |
Human | |
Untagged | |
10 mM sodium phosphate, pH 7.5 |