missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Inactive tyrosine-protein kinase 7 Protein
A DNA sequence encoding the Homo sapiens (Human) Inactive tyrosine-protein kinase 7, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP9763-ye-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001257327.1 | |
Inactive tyrosine-protein kinase 7 Protein | |
Research Use Only | |
PTK7 | |
Recombinant Protein | |
Yeast | |
AIVFIKQPSS QDALQGRRAL LRCEVEAPGP VHVYWLLDGA PVQDTERRFA QGSSLSFAAV DRLQDSGTFQ CVARDDVTGE EARSANASFN IKWIEAGPVV LKHPASEAEI QPQTQVTLRC HIDGHPRPTY QWFRDGTPLS DGQSNHTVSS KERNLTLRPA GPEHSGLYSC CAHSAFGQAC SSQNFTLSIA DESFARVVLA PQDVVVARYE EAMFHCQFSA QPPPSLQWLF EDETPITNRS RPPHLRRATV FANGSLLLTQ VRPRNAGIYR CIGQGQRGPP IILEATLHLA EIEDMPLFEP RVFTAGSEER VTCLPPKGLP EPSVWWEHAG VRLPTHGRVY QKGHELVLAN IAESDAGVYT CHAANLAGQR RQDVNITVAT VPSWLKKPQD SQLEEGKPGY LDCLTQATPK PTVVWYRNQM LISEDSRFEV FKNGTLRINS VEVYDGTWYR CMSSTPAGSI EAQARVQVLE KLKFTPPPQP QQCMEFDKEA TVPCSATGRE KPTIKWERAD GSSLPEWVTD NAGTLHFARV TRDDAGNYTC IASNGPQGQI RAHVQLTVAV FITFKVEPER TTVYQGHTAL LQCEAQGDPK PLIQWKGKDR ILDPTKLGPR MHIFQNGSLV IHDVAPEDSG RYTCIAGNSC NIKHTEAPLY VVDKPVPEES EGPGSPPPYK MIQT | |
Greater than 90% as determined by SDS-PAGE. |
5754 | |
50 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |