missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Integrin alpha-IIb Protein
A DNA sequence encoding the Homo sapiens (Human) Integrin alpha-IIb, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6239-ye-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_000410.2 | |
Integrin alpha-IIb Protein | |
Research Use Only | |
ITGA2B | |
Recombinant Protein | |
Yeast | |
CVPQLQLTAS VTGSPLLVGA DNVLELQMDA ANEGEGAYEA ELAVHLPQGA HYMRALSNVE GFERLICNQK KENETRVVLC ELGNPMKKNA QIGIAMLVSV GNLEEAGESV SFQLQIRSKN SQNPNSKIVL LDVPVRAEAQ VELRGNSFPA SLVVAAEEGE REQNSLDSWG PKVEHTYELH NNGPGTVNGL HLSIHLPGQS QPSDLLYILD IQPQGGLQCF PQPPVNPLKV DWGLPIPSPS PIHPAHHKR | |
Greater than 90% as determined by SDS-PAGE. |
3674 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |