missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Interferon alpha-21 Protein
A DNA sequence encoding the Homo sapiens (Human) Interferon alpha-21, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6201-ye-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_002166.2 | |
Interferon alpha-21 Protein | |
Research Use Only | |
IFNA21 | |
Recombinant Protein | |
Yeast | |
CDLPQTHSLG NRRALILLAQ MGRISPFSCL KDRHDFGFPQ EEFDGNQFQK AQAISVLHEM IQQTFNLFST KDSSATWEQS LLEKFSTELN QQLNDLEACV IQEVGVEETP LMNVDSILAV KKYFQRITLY LTEKKYSPCA WEVVRAEIMR SFSLSKIFQE RLRRKE | |
Greater than 90% as determined by SDS-PAGE. |
3452 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |