missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Keratin, type I cuticular Ha5 Protein
A DNA sequence encoding the Homo sapiens (Human) Keratin, type I cuticular Ha5, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7995-ec-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_002271.3 | |
Keratin, type I cuticular Ha5 Protein | |
Research Use Only | |
KRT35 | |
Recombinant Protein | |
E. coli | |
MASKCLKAGF SSGSLKSPGG ASGGSTRVSA MYSSSSCKLP SLSPVARSFS ACSVGLGRSS YRATSCLPAL CLPAGGFATS YSGGGGWFGE GILTGNEKET MQSLNDRLAG YLEKVRQLEQ ENASLESRIR EWCEQQVPYM CPDYQSYFRT IEELQKKTLC SKAENARLVV EIDNAKLAAD DFRTKYETEV SLRQLVESDI NGLRRILDDL TLCKSDLEAQ VESLKEELLC LKKNHEEEVN SLRCQLGDRL NVEVDAAPPV DLNRVLEEMR CQYETLVENN RRDAEDWLDT QSEELNQQVV SSSEQLQSCQ AEIIELRRTV NALEIELQAQ HSMRDALEST LAETEARYSS QLAQMQCMIT NVEAQLAEIR ADLERQNQEY QVLLDVRARL ECEINTYRGL LESEDSKLPC NPCAPDYSPS KSCLPCLPAA SCGPSAARTN CSPRPICVPC PGGRF | |
Greater than 90% as determined by SDS-PAGE. |
3886 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |