missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Keratin, type II cytoskeletal 4 Protein
A DNA sequence encoding the Homo sapiens (Human) Keratin, type II cytoskeletal 4, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6281-ye-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
Keratin, type II cytoskeletal 4 Protein | |
Research Use Only | |
KRT4 | |
Recombinant Protein | |
Yeast | |
MIARQQCVRG GPRGFSCGSA IVGGGKRGAF SSVSMSGGAG RCSSGGFGSR SLYNLRGNKS ISMSVAGSRQ GACFGGAGGF GTGGFGAGGF GAGFGTGGFG GGFGGSFSGK GGPGFPVCPA GGIQEVTINQ SLLTPLHVEI DPEIQKVRTE EREQIKLLNN KFASFIDKVQ FLEQQNKVLE TKWNLLQQQT TTTSSKNLEP LFETYLSVLR KQLDTLGNDK GRLQSELKTM QDSVEDFKTK YEEEINKRTA AENDFVVLKK DVDAAYLNKV ELEAKVDSLN DEINFLKVLY DAELSQMQTH VSDTSVVLSM DNNRNLDLDS IIAEVRAQYE EIAQRSKAEA EALYQTKVQQ LQISVDQHGD NLKNTKSEIA ELNRMIQRLR AEIENIKKQC QTLQVSVADA EQRGENALKD AHSKRVELEA ALQQAKEELA RMLREYQELM SVKLALDIEI ATYRKLLEGE EYRMSGECQS AVSISVVSGS TSTGGISGGL GSGSGFGLSS GFGSGSGSGF GFGGSVSGSS SSKIISTTTL NKRR | |
Greater than 90% as determined by SDS-PAGE. |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |