missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human KLK7 / PRSS6 Protein
A DNA sequence encoding the Homo sapiens (Human) KLK7 / PRSS6, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6267-ec-200ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001193982.1 | |
KLK7 / PRSS6 Protein | |
Research Use Only | |
KLK7 | |
Recombinant Protein | |
E. coli | |
DKIIDGAPCA RGSHPWQVAL LSGNQLHCGG VLVNERWVLT AAHCKMNEYT VHLGSDTLGD RRAQRIKASK SFRHPGYSTQ THVNDLMLVK LNSQARLSSM VKKVRLPSRC EPPGTTCTVS GWGTTTSPDV TFPSDLMCVD VKLISPQDCT KVYKDLLENS MLCAGIPDSK KNACNGDSGG PLVCRGTLQG LVSWGTFPCG QPNDPGVYTQ VCKFTKWIND TMKKHR | |
Greater than 90% as determined by SDS-PAGE. |
5650 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |